missing translation for 'onlineSavingsMsg'
Learn More

Nepsilon-Boc-L-lysine tert-butyl ester hydrochloride, 95%

Catalog No. p-7054625
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6299303 1 g
AAH6299306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6299303 Supplier Thermo Scientific Chemicals Supplier No. H6299303
Only null left

CAS: 13288-57-8 | C15H31ClN2O4 | 338.87 g/mol

Nepsilon-Boc-L-lysine tert-butyl ester hydrochloride is an pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nepsilon-Boc-L-lysine tert-butyl ester hydrochloride is an pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 13288-57-8
Molecular Formula C15H31ClN2O4
Molecular Weight (g/mol) 338.87
MDL Number MFCD00038899
InChI Key TZBPQINFXPIRBX-VAMKTSTMNA-N
Synonym h-lys boc-otbu.hcl, h-lys boc-otbu hcl, ne-boc-l-lysine tert-butyl ester hydrochloride, h-l-lys boc-otbu*hcl, c15h30n2o4.hcl, pubchem18975, lys boc-otbu hcl, h-lys boc-obut hcl, h-lys boc-otbu?hcl, h-lys boc-otbu hydrochloride
PubChem CID 13819885
SMILES Cl.CC(C)(C)OC(=O)NCCCC[C@H](N)C(=O)OC(C)(C)C

Specifications

Color White
Solubility Information Insoluble in water.
IUPAC Name tert-butyl (2S)-2-amino-6-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoate;hydrochloride
Quantity 1 g
Formula Weight 338.87
Percent Purity 95%
Physical Form Powder
Chemical Name or Material Nepsilon-Boc-L-lysine tert-butyl ester hydrochloride
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only