missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nepsilon-Boc-L-lysine tert-butyl ester hydrochloride, 95%
Description
Nepsilon-Boc-L-lysine tert-butyl ester hydrochloride is an pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Nepsilon-Boc-L-lysine tert-butyl ester hydrochloride is an pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Nepsilon-Boc-L-lysine tert-butyl ester hydrochloride is an pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Specifications
Specifications
| Color | White |
| Solubility Information | Insoluble in water. |
| IUPAC Name | tert-butyl 2-amino-6-{[(tert-butoxy)carbonyl]amino}hexanoate hydrochloride |
| Quantity | 5 g |
| Formula Weight | 338.87 |
| Percent Purity | 95% |
| Physical Form | Powder |
| Chemical Name or Material | Nepsilon-Boc-L-lysine tert-butyl ester hydrochloride |
Safety and Handling
| TSCA | No |
| storageNote1 | -30° to -10°C |
| Recommended Storage | Store at -20°C |
RUO – Research Use Only