missing translation for 'onlineSavingsMsg'
Learn More

Nepsilon-Allyloxycarbonyl-Nalpha-Fmoc-D-lysine, 95%

Catalog No. AAH6686106
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6686106 5 g
AAH6686103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6686106 Supplier Thermo Scientific Chemicals Supplier No. H6686106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 214750-75-1 | C25H28N2O6 | 452.51 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 214750-75-1
Molecular Formula C25H28N2O6
Molecular Weight (g/mol) 452.51
MDL Number MFCD00798639
InChI Key OJBNDXHENJDCBA-JOCHJYFZSA-N
Synonym fmoc-d-lys alloc-oh, fmoc-d-lys aloc-oh, n-fmoc-n'-allyloxycarbonyl-d-lysine, r-2-9h-fluoren-9-yl methoxy carbonyl amino-6-allyloxy carbonyl amino hexanoic acid, ambotzfaa1329, n, a-alloc-n, a-fmoc-d-lysine, a-fmoc-n, a-alloc-d-lysine, fmoc-d-lys alloc-oh hplc, n-alpha-9-fluorenylmethyloxycarbonyl-n-epsilon-allyl-oxycarbonyl-d-lysine, n∼2∼-9h-fluoren-9-yl methoxy carbonyl-n∼6∼-prop-2-en-1-yl oxy carbonyl-d-lysine
PubChem CID 51340505
IUPAC Name (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-6-(prop-2-enoxycarbonylamino)hexanoic acid
SMILES OC(=O)[C@@H](CCCCNC(=O)OCC=C)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Color White
Quantity 5 g
Formula Weight 452.51
Percent Purity 95%
Physical Form Powder
Chemical Name or Material Nepsilon-Allyloxycarbonyl-Nalpha-Fmoc-D-lysine
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only