missing translation for 'onlineSavingsMsg'
Learn More

Nepsilon-Acetyl-Nalpha-Boc-L-lysine, 98%, Thermo Scientific™

Catalog No. p-7041499
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6234706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6234706 Supplier Thermo Scientific Chemicals Supplier No. H6234706

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 6404-26-8
Molecular Formula C13H24N2O5
Molecular Weight (g/mol) 288.34
MDL Number MFCD00057791
InChI Key IOKOUUAPSRCSNT-UEQNJFAPNA-N
Synonym boc-lys ac-oh, n-boc-n'-acetyl-l-lysine, s-6-acetamido-2-tert-butoxycarbonyl amino hexanoic acid, boc-lys-ac-oh, n-boc-n-acetyl-l-lisine, boc-n-epsilon-acetyl-l-lysine, na-t-boc-n-epsilon-acetyl-l-lysine, 2s-6-acetylamino-2-tert-butoxycarbonylamino hexanoic acid, 2s-2-tert-butoxycarbonyl amino-6-acetamidohexanoic acid
PubChem CID 7016049
IUPAC Name (2S)-6-acetamido-2-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid
SMILES CC(=O)NCCCC[C@H](NC(=O)OC(C)(C)C)C(O)=O

Specifications

Quantity 5 g
Formula Weight 288.34
Percent Purity 98%
Chemical Name or Material Nepsilon-Acetyl-Nalpha-Boc-L-lysine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold