missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nepsilon-4-[4-(Dimethylamino)phenylazo]benzoyl-Nalpha-Fmoc-L-lysine, 95%
CAS: 146998-27-8 | C36H37N5O5 | 619.72 g/mol
$227.73 - $741.42
Chemical Identifiers
| CAS | 146998-27-8 |
|---|---|
| Molecular Formula | C36H37N5O5 |
| Molecular Weight (g/mol) | 619.72 |
| MDL Number | MFCD00467694 |
| InChI Key | FPOPWTDBGMLRNG-YGFIFORBNA-N |
| Synonym | fmoc-lys dabcyl-oh, a-4-4-dimethylamino phenylazo benzoyl-n, s,e-2-9h-fluoren-9-yl methoxy carbonyl amino-6-4-4-dimethylamino phenyl diazenyl benzamido hexanoic acid, ambotzfaa1498, s,e-2-9h-fluoren-9-yl methoxy carbonyl amino-6-4-4-dimethylamino phenyl diazenyl benzamido hexanoic acid, n, fmoc-l-lys dabcyl-oh, a-dabcyl-l-lysine, a-fmoc-n, a-dabcyl-l-lysine, nalpha-fmoc-nepsilon-dabcyl-l-lysine, na-fmoc-ne-dimethylaminophenyldiazobenzoyl-l-lysine |
| PubChem CID | 2756070 |
| IUPAC Name | (2S)-6-[[4-[[4-(dimethylamino)phenyl]diazenyl]benzoyl]amino]-2-(9H-fluoren-9-ylmethoxycarbonylamino)hexanoic acid |
| SMILES | CN(C)C1=CC=C(C=C1)N=NC1=CC=C(C=C1)C(=O)NCCCC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH63214MD
|
Thermo Scientific Chemicals
H63214MD |
250 mg |
Each for $227.73
|
|
|||||
|
AAH6321403
|
Thermo Scientific Chemicals
H6321403 |
1 g |
Each for $741.42
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 146998-27-8 | |
| 619.72 | |
| FPOPWTDBGMLRNG-YGFIFORBNA-N | |
| 2756070 | |
| CN(C)C1=CC=C(C=C1)N=NC1=CC=C(C=C1)C(=O)NCCCC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O |
| C36H37N5O5 | |
| MFCD00467694 | |
| fmoc-lys dabcyl-oh, a-4-4-dimethylamino phenylazo benzoyl-n, s,e-2-9h-fluoren-9-yl methoxy carbonyl amino-6-4-4-dimethylamino phenyl diazenyl benzamido hexanoic acid, ambotzfaa1498, s,e-2-9h-fluoren-9-yl methoxy carbonyl amino-6-4-4-dimethylamino phenyl diazenyl benzamido hexanoic acid, n, fmoc-l-lys dabcyl-oh, a-dabcyl-l-lysine, a-fmoc-n, a-dabcyl-l-lysine, nalpha-fmoc-nepsilon-dabcyl-l-lysine, na-fmoc-ne-dimethylaminophenyldiazobenzoyl-l-lysine | |
| (2S)-6-[[4-[[4-(dimethylamino)phenyl]diazenyl]benzoyl]amino]-2-(9H-fluoren-9-ylmethoxycarbonylamino)hexanoic acid |
Specifications
| 146998-27-8 | |
| C36H37N5O5 | |
| fmoc-lys dabcyl-oh, a-4-4-dimethylamino phenylazo benzoyl-n, s,e-2-9h-fluoren-9-yl methoxy carbonyl amino-6-4-4-dimethylamino phenyl diazenyl benzamido hexanoic acid, ambotzfaa1498, s,e-2-9h-fluoren-9-yl methoxy carbonyl amino-6-4-4-dimethylamino phenyl diazenyl benzamido hexanoic acid, n, fmoc-l-lys dabcyl-oh, a-dabcyl-l-lysine, a-fmoc-n, a-dabcyl-l-lysine, nalpha-fmoc-nepsilon-dabcyl-l-lysine, na-fmoc-ne-dimethylaminophenyldiazobenzoyl-l-lysine | |
| CN(C)C1=CC=C(C=C1)N=NC1=CC=C(C=C1)C(=O)NCCCC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O | |
| 250 mg | |
| 2756070 | |
| 95% | |
| Nepsilon-4-[4-(Dimethylamino)phenylazo]benzoyl-Nalpha-Fmoc-L-lysine |
| Red | |
| MFCD00467694 | |
| FPOPWTDBGMLRNG-YGFIFORBNA-N | |
| (2S)-6-[[4-[[4-(dimethylamino)phenyl]diazenyl]benzoyl]amino]-2-(9H-fluoren-9-ylmethoxycarbonylamino)hexanoic acid | |
| 619.72 | |
| 619.71 | |
| Crystalline Powder |
Safety and Handling
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only