missing translation for 'onlineSavingsMsg'
Learn More

Nepsilon-4-[4-(Dimethylamino)phenylazo]benzoyl-Nalpha-Fmoc-L-lysine, 95%

Catalog No. AAH63214MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH63214MD 250 mg
AAH6321403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH63214MD Supplier Thermo Scientific Chemicals Supplier No. H63214MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 146998-27-8 | C36H37N5O5 | 619.72 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 146998-27-8
Molecular Formula C36H37N5O5
Molecular Weight (g/mol) 619.72
MDL Number MFCD00467694
InChI Key FPOPWTDBGMLRNG-YGFIFORBNA-N
Synonym fmoc-lys dabcyl-oh, fmoc-lys dadcyl-oh, s,e-2-9h-fluoren-9-yl methoxy carbonyl amino-6-4-4-dimethylamino phenyl diazenyl benzamido hexanoic acid, ambotzfaa1498, fmoc-l-lys dabcyl-oh, n, a-fmoc-n, a-dabcyl-l-lysine, nalpha-fmoc-nepsilon-dabcyl-l-lysine, na-fmoc-ne-dimethylaminophenyldiazobenzoyl-l-lysine, a-4-4-dimethylamino phenylazo benzoyl-n, a-fmoc-l-lysine
PubChem CID 2756070
IUPAC Name (2S)-6-[[4-[[4-(dimethylamino)phenyl]diazenyl]benzoyl]amino]-2-(9H-fluoren-9-ylmethoxycarbonylamino)hexanoic acid
SMILES CN(C)C1=CC=C(C=C1)N=NC1=CC=C(C=C1)C(=O)NCCCC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Specifications

Color Red
Quantity 250 mg
Formula Weight 619.71
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material Nepsilon-4-[4-(Dimethylamino)phenylazo]benzoyl-Nalpha-Fmoc-L-lysine
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only