missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neopentylamine 98.0+%, TCI America™
Supplier: TCI America N050525ML
Specifications
| Neopentylamine | |
| Colorless | |
| C5H13N | |
| 25 mL | |
| neopentylamine, 2,2-dimethylpropylamine, 1-propanamine, 2,2-dimethyl, 2,2-dimethyl-1-propylamine, 1-amino-2,2-dimethylpropane, neo-c5h11nh2, 2,2-dimethyl-1-propylamin, neo-pentylamine, neopentyl amine, 2,2-dimethylpropanamine | |
| CC(C)(C)CN | |
| 87.17 | |
| 87.17 | |
| Liquid |
| 5813-64-9 | |
| 82°C | |
| MFCD00008134 | |
| 2733 | |
| XDIAMRVROCPPBK-UHFFFAOYSA-N | |
| 2,2-dimethylpropan-1-amine | |
| 79882 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 5813-64-9 | |
| 87.17 | |
| XDIAMRVROCPPBK-UHFFFAOYSA-N | |
| 79882 | |
| CC(C)(C)CN |
| C5H13N | |
| MFCD00008134 | |
| neopentylamine, 2,2-dimethylpropylamine, 1-propanamine, 2,2-dimethyl, 2,2-dimethyl-1-propylamine, 1-amino-2,2-dimethylpropane, neo-c5h11nh2, 2,2-dimethyl-1-propylamin, neo-pentylamine, neopentyl amine, 2,2-dimethylpropanamine | |
| 2,2-dimethylpropan-1-amine |
Safety and Handling
EINECSNumber : (2)-0133
TSCA : No