missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neopentyl Glycol Mono(hydroxypivalate) 97.0+%, TCI America™
$63.52 - $84.70
Chemical Identifiers
| CAS | 1115-20-4 |
|---|---|
| Molecular Formula | C10H20O4 |
| Molecular Weight (g/mol) | 204.266 |
| MDL Number | MFCD00059597 |
| InChI Key | SZCWBURCISJFEZ-UHFFFAOYSA-N |
| Synonym | hydroxypivalic acid neopentyl glycol ester, esterdiol 204, 3-hydroxy-2,2-dimethylpropyl 3-hydroxy-2,2-dimethylpropionate, hydroxyneopentyl hydroxypivalate, unii-cg0a37gahj, propanoic acid, 3-hydroxy-2,2-dimethyl-, 3-hydroxy-2,2-dimethylpropyl ester, neopentyl glycol monohydroxypivalate, 3-hydroxypivaloyloxy-2,2-dimethylpropanol, 3-hydroxy-2,2-dimethylpropyl hydroxypivalate, neopental glycol monohydroxypivalate |
| PubChem CID | 14218 |
| IUPAC Name | (3-hydroxy-2,2-dimethylpropyl) 3-hydroxy-2,2-dimethylpropanoate |
| SMILES | CC(C)(CO)COC(=O)C(C)(C)CO |
Chemical Identifiers
| 1115-20-4 | |
| 204.266 | |
| SZCWBURCISJFEZ-UHFFFAOYSA-N | |
| 14218 | |
| CC(C)(CO)COC(=O)C(C)(C)CO |
| C10H20O4 | |
| MFCD00059597 | |
| hydroxypivalic acid neopentyl glycol ester, esterdiol 204, 3-hydroxy-2,2-dimethylpropyl 3-hydroxy-2,2-dimethylpropionate, hydroxyneopentyl hydroxypivalate, unii-cg0a37gahj, propanoic acid, 3-hydroxy-2,2-dimethyl-, 3-hydroxy-2,2-dimethylpropyl ester, neopentyl glycol monohydroxypivalate, 3-hydroxypivaloyloxy-2,2-dimethylpropanol, 3-hydroxy-2,2-dimethylpropyl hydroxypivalate, neopental glycol monohydroxypivalate | |
| (3-hydroxy-2,2-dimethylpropyl) 3-hydroxy-2,2-dimethylpropanoate |
Specifications
| 1115-20-4 | |
| White | |
| C10H20O4 | |
| 25 g | |
| SZCWBURCISJFEZ-UHFFFAOYSA-N | |
| (3-hydroxy-2,2-dimethylpropyl) 3-hydroxy-2,2-dimethylpropanoate | |
| 14218 | |
| ≥97.0% (GC) | |
| Neopentyl Glycol Mono(hydroxypivalate) |
| 51°C | |
| 292°C | |
| MFCD00059597 | |
| hydroxypivalic acid neopentyl glycol ester, esterdiol 204, 3-hydroxy-2,2-dimethylpropyl 3-hydroxy-2,2-dimethylpropionate, hydroxyneopentyl hydroxypivalate, unii-cg0a37gahj, propanoic acid, 3-hydroxy-2,2-dimethyl-, 3-hydroxy-2,2-dimethylpropyl ester, neopentyl glycol monohydroxypivalate, 3-hydroxypivaloyloxy-2,2-dimethylpropanol, 3-hydroxy-2,2-dimethylpropyl hydroxypivalate, neopental glycol monohydroxypivalate | |
| CC(C)(CO)COC(=O)C(C)(C)CO | |
| 204.266 | |
| 204.27 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (2)-1353&(2)-1406&(2)-2733
TSCA : Yes