missing translation for 'onlineSavingsMsg'
Learn More

Neopentyl Glycol Mono(hydroxypivalate) 97.0+%, TCI America™

Catalog No. N0461500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0461500G 500 g
N046125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N0461500G Supplier TCI America Supplier No. N0461500G

Chemical Identifiers

CAS 1115-20-4
Molecular Formula C10H20O4
Molecular Weight (g/mol) 204.266
MDL Number MFCD00059597
InChI Key SZCWBURCISJFEZ-UHFFFAOYSA-N
Synonym hydroxypivalic acid neopentyl glycol ester, esterdiol 204, 3-hydroxy-2,2-dimethylpropyl 3-hydroxy-2,2-dimethylpropionate, hydroxyneopentyl hydroxypivalate, unii-cg0a37gahj, propanoic acid, 3-hydroxy-2,2-dimethyl-, 3-hydroxy-2,2-dimethylpropyl ester, neopentyl glycol monohydroxypivalate, 3-hydroxypivaloyloxy-2,2-dimethylpropanol, 3-hydroxy-2,2-dimethylpropyl hydroxypivalate, neopental glycol monohydroxypivalate
PubChem CID 14218
IUPAC Name (3-hydroxy-2,2-dimethylpropyl) 3-hydroxy-2,2-dimethylpropanoate
SMILES CC(C)(CO)COC(=O)C(C)(C)CO

Specifications

Melting Point 51°C
Color White
Boiling Point 292°C
Quantity 500 g
Formula Weight 204.27
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Neopentyl Glycol Mono(hydroxypivalate)
EINECSNumber (2)-1353&(2)-1406&(2)-2733
TSCA Yes