missing translation for 'onlineSavingsMsg'
Learn More

Neopentyl Glycol Dimethacrylate (stabilized with MEHQ) 90.0+%, TCI America™

Catalog No. N0780500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0780500G 500 g
N078025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N0780500G Supplier TCI America Supplier No. N0780500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1985-51-9
Molecular Formula C13H20O4
Molecular Weight (g/mol) 240.30
MDL Number MFCD00048120
InChI Key ULQMPOIOSDXIGC-UHFFFAOYSA-N
Synonym 1,3-Bis(methacryloyloxy)-2,2-dimethylpropane, 2,2-Dimethyl-1,3-propanediol Dimethacrylate
PubChem CID 16135
IUPAC Name 2,2-dimethyl-3-[(2-methylprop-2-enoyl)oxy]propyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCC(C)(C)COC(=O)C(C)=C

Specifications

Color Colorless
Quantity 500 g
Formula Weight 240.30
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material Neopentyl Glycol Dimethacrylate (stabilized with MEHQ)
EINECSNumber (2)-0958&(2)-1059&(2)-1062
TSCA Yes