Learn More
Invitrogen™ Neon™ NxT Electroporation System 8-Channel Tubes
Catalog No. NT8
Change view
Click to view available options
Quantity:
8 Tubes
Catalog No. Quantity
NT8 8 Tubes
1 options
Catalog No. NT8 Supplier Invitrogen™ Supplier No. NT8
Season of Savings
View all promotions
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Neon NxT Electroporation System 8-Channel Tubes are designed specifically for use with the Neon NxT Electroporation System with 8-Channel Pipette.
Neon NxT Electroporation System 8-Channel Tubes are designed specifically for use with the Neon NxT Electroporation System with 8-Channel Pipette (Cat. Nos. NEON18S, NEON18SK, NEON8, NEON8U). Each Neon NxT Electroporation System Kit with 8-Channel Tubes for 96 x 2 reactions (Cat. Nos. N1096-8 and N10096-8) comes with four 8-channel tubes and each Neon NxT Electroporation System Kit for 384 x 2 reactions (Cat. Nos. N10384-8 and N100384-8) comes with sixteen 8-channel tubes. This pack of eight tubes is offered for those wishing to use one tube per eight reactions (tubes can be used up to twelve times) depending on their experimental setup.
Specifications
Includes Includes 8 x 8-chamber tubes, use at room temperature under normal laboratory conditions. Refer to the Neon™ NxT Electroporation System user manual for protocol instructions.
Quantity 8 Tubes
For Use With (Equipment) Neon NxT Electroporation System
For Use With (Application) Electroporation
For Research Use Only. Not for use in diagnostic procedures.