missing translation for 'onlineSavingsMsg'
Learn More

Neomycin sulfate hydrate

Catalog No. AAJ6149922
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6149922 100 g
AAJ6149914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6149922 Supplier Thermo Scientific Chemicals Supplier No. J6149922

CAS: 1405-10-3 | C23H48N6O17S | 712.72 g/mol

Neomycin sulfate is an aminoglycoside antibiotic and calcium channel protein inhibitor. Neomycin sulfate also binds to prokaryotic ribosomes inhibiting translation and is effective against gram-positive and gram-negative bacteria. Neomycin sulfate inhibits Phospholipase C via binding to inositol phospholipids. It also inhibits phosphatidylcholine-PLD activity and induces Ca2+ mobilization and PLA2 activation in human platelets. Neomycin sulfate inhibits DNase I induced DNA degradation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Neomycin sulfate is an aminoglycoside antibiotic and calcium channel protein inhibitor. Neomycin sulfate also binds to prokaryotic ribosomes inhibiting translation and is effective against gram-positive and gram-negative bacteria. Neomycin sulfate inhibits Phospholipase C via binding to inositol phospholipids. It also inhibits phosphatidylcholine-PLD activity and induces Ca2+ mobilization and PLA2 activation in human platelets. Neomycin sulfate inhibits DNase I induced DNA degradation.

Solubility
Soluble in water

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.

Chemical Identifiers

CAS 1405-10-3
Molecular Formula C23H48N6O17S
Molecular Weight (g/mol) 712.72
MDL Number MFCD29905465
InChI Key OIXVKQDWLFHVGR-DJUBFIBWNA-N
PubChem CID 124080932
SMILES OS(O)(=O)=O.NCC1OC(O[C@@H]2[C@@H](CO)O[C@@H](OC3C(O)C(N)CC(N)C3OC3OC(CN)C(O)C(O)C3N)[C@@H]2O)C(N)C(O)C1O

Specifications

Type Neomycin Sulfate
Physical Form Powder
Color White
Quantity 100 g
Merck Index 14,6454
Solubility Information Soluble in water
IUPAC Name (2S,3R,4S,5S,6R)-5-amino-2-(aminomethyl)-6-[(1R,2R,3S,4R,6R)-4,6-diamino-2-[(2R,3S,4R,5S)-4-[(2S,3R,4S,5R,6S)-3-amino-6-(aminomethyl)-4,5-dihydroxyoxan-2-yl]oxy-3-hydroxy-5-(hydroxymethyl)oxolan-2-yl]oxy-3-hydroxycyclohexyl]oxyoxane-3,4-diol;sulfuric acid
Formula Weight 908.87 (Anhydrous)
Chemical Name or Material Neomycin sulfate hydrate
Hazard Category H317-H334-H361d-H373
Hazard Statement GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P260-P272-P280g-P281-P285-P302+P352-P304+P341-P308+P313-P333+P313-P342+P311-P363-P501c
EINECSNumber 215-773-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only