missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(+)-Neomenthol 96.0+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Boiling Point | 212°C |
| Quantity | 25 mL |
| Formula Weight | 156.27 |
| Percent Purity | ≥96.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | (+)-Neomenthol |
Safety and Handling
| EINECSNumber | (3)-2333 |
| TSCA | Yes |