missing translation for 'onlineSavingsMsg'
Learn More

(+)-Neomenthol 96.0+%, TCI America™

Catalog No. N06265ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N06265ML 5 mL
N062625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N06265ML Supplier TCI America Supplier No. N06265ML
Only null left

Chemical Identifiers

CAS 2216-52-6
Molecular Formula C10H20O
Molecular Weight (g/mol) 156.269
MDL Number MFCD00062980
InChI Key NOOLISFMXDJSKH-UTLUCORTSA-N
Synonym (1S,2S,5R)-2-Isopropyl-5-methylcyclohexanol
PubChem CID 439263
ChEBI CHEBI:15402
IUPAC Name (1S,2S,5R)-5-methyl-2-propan-2-ylcyclohexan-1-ol
SMILES CC1CCC(C(C1)O)C(C)C

Specifications

Color Colorless
Boiling Point 212°C
Quantity 5 mL
Formula Weight 156.27
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material (+)-Neomenthol
EINECSNumber (3)-2333
TSCA Yes