missing translation for 'onlineSavingsMsg'
Learn More

Neohesperidin Dihydrochalcone Hydrate 98.0+%, TCI America™

Catalog No. p-7133928
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N06755G 5 g
N067525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N06755G Supplier TCI America Supplier No. N06755G
Only null left

Chemical Identifiers

CAS 20702-77-6
Molecular Formula C28H36O15
Molecular Weight (g/mol) 612.58
MDL Number MFCD00017711
InChI Key ITVGXXMINPYUHD-UHFFFAOYSA-N
Synonym neohesperidin dihydrochalcone, nhdc, neohesperidin dc, neohesperidin dhc, unii-3x476d83qv, ccris 4848, neosperidin dihydrochalcone, neohesperidine dihydrochalcone, 1-4-2-o-6-deoxy-alpha-l-mannopyranosyl-beta-d-glucopyranosyl oxy-2,6-dihydroxyphenyl-3-3-hydroxy-4-methoxyphenyl propan-1-one, 1-4-2-o-6-deoxy-alpha-l-mannopyranosyl-beta-d-glucopyranosyl oxy-2,6-dihydroxyphenyl-3-3-hydroxy-4-methoxyphenyl
PubChem CID 30231
ChEBI CHEBI:83535
IUPAC Name 1-(4-{[4,5-dihydroxy-6-(hydroxymethyl)-3-[(3,4,5-trihydroxy-6-methyloxan-2-yl)oxy]oxan-2-yl]oxy}-2,6-dihydroxyphenyl)-3-(3-hydroxy-4-methoxyphenyl)propan-1-one
SMILES COC1=C(O)C=C(CCC(=O)C2=C(O)C=C(OC3OC(CO)C(O)C(O)C3OC3OC(C)C(O)C(O)C3O)C=C2O)C=C1

Specifications

Quantity 5 g
Formula Weight 612.58
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Neohesperidin Dihydrochalcone Hydrate
EINECSNumber (4)-1875
RTECSNumber LZ5785000
TSCA Yes