missing translation for 'onlineSavingsMsg'
Learn More

ICU Medical Neoheel™ Safety Lancets

Catalog No. 22024993
Encompass
Change view
Click to view available options
Quantity:
50/Box
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22024993 50/Box
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22024993 Supplier ICU Medical Supplier No. 1052N

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Neoheel™ Safety Lancets for collecting quality blood samples from newborns

  • Used to collect quality blood samples from infants for metabolic screening as well as glucose and other tests
  • Automatic retraction is designed to avoid needlestick injuries and cross-contamination exposure
  • The safety lancet produces less bruising and requires less punctures to obtain sufficient volumes of blood
  • Arc shaped design reduces the number of heel puncture sites, time taken to complete the test and time the baby cries
  • Integral safety tab prevents premature activation while avoiding additional loose parts

Specifications

Disposable Single-use
Product Type Safety Lancet
Color Yellow
For Use With (Application) Newborn Blood Collection
Quantity 50/Box