missing translation for 'onlineSavingsMsg'
Learn More

Neodymium powder, -40 mesh, 99.8% (REO)

Catalog No. p-7059211
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4418106 5 g
AA4418114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4418106 Supplier Thermo Scientific Chemicals Supplier No. 04418106
Only null left

CAS: 7440-00-8 | Nd | 144.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon

Chemical Identifiers

CAS 7440-00-8
Molecular Formula Nd
Molecular Weight (g/mol) 144.24
MDL Number MFCD00011130
InChI Key QEFYFXOXNSNQGX-UHFFFAOYSA-N
Synonym neodimio, neodyme, neodym, trihydride, ingot reo, neodymium, ion nd5+, atom, ingot, iii, foil, 3n
PubChem CID 23934
ChEBI CHEBI:33372
IUPAC Name neodymium
SMILES [Nd]

Specifications

Color Gray to Silver
Quantity 5 g
Mesh Size −40 Mesh
Odor Odorless
Packaging Ampule
Sensitivity Air and moisture sensitive
Form Powder
UN Number UN3089
Percent Purity 99.9%
Assay (REO)
Chemical Name or Material Neodymium powder
Show More Show Less
Hazard Category H228-H315-H319-X
Hazard Statement GHS H Statement
H228-H260
Flammable solid.
In contact with water releases flammable gases which may ignite spontaneously.
Precautionary Statement P210-P235+P410-P240-P241-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q
DOTInformation Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.
EINECSNumber 231-109-3
RTECSNumber QO8575000
TSCA Yes
storageNote1
  • Ampouled under argon
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only