missing translation for 'onlineSavingsMsg'
Learn More

Neodymium ingot, 99.1% (metals basis)

Catalog No. p-7048201
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA0021414 25 g
AA0021422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA0021414 Supplier Thermo Scientific Chemicals Supplier No. 00021414
Only null left

CAS: 7440-00-8 | Nd | 144.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under mineral oil

Chemical Identifiers

CAS 7440-00-8
Molecular Formula Nd
Molecular Weight (g/mol) 144.24
MDL Number MFCD00011130
InChI Key QEFYFXOXNSNQGX-UHFFFAOYSA-N
Synonym neodimio, neodyme, neodym, trihydride, ingot reo, neodymium, ion nd5+, atom, ingot, iii, foil, 3n
PubChem CID 23934
ChEBI CHEBI:33372
IUPAC Name neodymium
SMILES [Nd]

Specifications

Color Gray-Silver
Quantity 25 g
Form Ingot
Percent Purity 99.1%
Assay (metals basis)
Chemical Name or Material Neodymium ingot
EINECSNumber 231-109-3
RTECSNumber QO8575000
TSCA Yes
storageNote1
  • Packaged under mineral oil
Recommended Storage Ambient temperatures

RUO – Research Use Only