missing translation for 'onlineSavingsMsg'
Learn More

Neodymium(III) sulfate octahydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

$596.67 - $596.67

Chemical Identifiers

CAS 13477-91-3
MDL Number MFCD00149803
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1318514
View Documents
Thermo Scientific Chemicals
01318514
25 g
Each for $596.67
Only null left
 
Description

Description

Neodymium(III) sulfate is used as catalyst, glass, crystal and capacitors, in CRT displays to enhance contrast between reds and greens.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13477-91-3
MFCD00149803

Specifications

1176°C
Crystalline
MFCD00149803
Soluble in water (80g/L) at 20°C moderately soluble in strong mineral acids.
99.99%
Odorless
Neodymium(III) sulfate octahydrate
13477-91-3
25 g
14,6450
720.78 (576.66 Anhydrous)
Reacton
2.85 g/mL
(REO)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 233-262-1

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only