missing translation for 'onlineSavingsMsg'
Learn More

Neodymium(III) sulfate octahydrate, 99.9% (REO)

Catalog No. p-4782012
Change view
Click to view available options
Quantity:
100 g
25 g
Catalog No. Quantity
AA4025314 25 g
AA4025322 100 g
2 options

Catalog No. AA4025314

Supplier: Thermo Scientific Chemicals 04025314

Only null left

CAS: 13477-91-3 | Nd2(SO4)3·8H2O

It is used as catalyst, glass, crystal and capacitors, in CRT displays to enhance contrast between reds and greens.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13477-91-3
Molecular Formula Nd2(SO4)3·8H2O
MDL Number MFCD00149803

Specifications

Melting Point 1176°C
Color Pink
Physical Form Crystals
Quantity 25 g
Assay Percent Range 99.9%
Solubility Information Soluble in water
Formula Weight 720.78 (576.66 Anhydrous)
Odor Odorless
Appearance Pink crystals
Density 2.85 g/cm3 at 20°C
Chemical Name or Material Neodymium(III) sulfate octahydrate, Rare Earth Oxides Content
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 233-262-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only