missing translation for 'onlineSavingsMsg'
Learn More

Neodymium(III) sulfate octahydrate, 99.9% (REO)

Catalog No. AA4025322
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4025322 100 g
AA4025314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4025322 Supplier Thermo Scientific Chemicals Supplier No. 04025322

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13477-91-3 | Nd2(SO4)3·8H2O

It is used as catalyst, glass, crystal and capacitors, in CRT displays to enhance contrast between reds and greens.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13477-91-3
Molecular Formula Nd2(SO4)3·8H2O
MDL Number MFCD00149803

Specifications

Melting Point 1176°C
Color Pink
Physical Form Crystals
Quantity 100 g
Assay Percent Range 99.9%
Solubility Information Soluble in water
Formula Weight 720.78 (576.66 Anhydrous)
Odor Odorless
Appearance Pink crystals
Density 2.85
Chemical Name or Material Neodymium(III) sulfate octahydrate, Rare Earth Oxides Content
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 233-262-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only