missing translation for 'onlineSavingsMsg'
Learn More

Neodymium(III) oxide, 99.9% (REO)

Catalog No. p-4408133
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3627514 25 g
AA3627522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3627514 Supplier Thermo Scientific Chemicals Supplier No. 03627514
Only null left

CAS: 1313-97-9 | Nd2O3 | 336.48 g/mol

Neodymium is used in coloring glasses and for doping some glass lasers since it transmits 90% of the blue, green, and red light rays. Neodymium compounds are used in glass coloring, permanent magnets, capacitors, infrared filters, resonators, Catalysis (diene polymerization), laser materials. Neodymium(III) oxide is used to dope glass, including sunglasses, to make solid-state lasers, and to color glasses and enamels.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1313-97-9
Molecular Formula Nd2O3
Molecular Weight (g/mol) 336.48
MDL Number MFCD00011134
InChI Key PLDDOISOJJCEMH-UHFFFAOYSA-N
IUPAC Name dineodymium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Nd+3].[Nd+3]

Specifications

Melting Point 2272°C
Color Blue
Physical Form Sintered Lump
Quantity 25 g
Assay Percent Range 99.9%
Solubility Information Insoluble in water. Soluble in mineral acids.
Formula Weight 336.48
Appearance Light blue 3-12mm Sintered Lump
Density 7.24 g/cm3 at 20°C
Chemical Name or Material Neodymium(III) oxide
EINECSNumber 215-214-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only