missing translation for 'onlineSavingsMsg'
Learn More

Neodymium(III) nitride, 99.9% (REO)

Catalog No. p-7051837
Change view
Click to view available options
Quantity:
10 g
2 g
Catalog No. Quantity
AA4209104 2 g
AA4209109 10 g
2 options

Catalog No. AA4209104

Supplier: Thermo Scientific Chemicals 04209104

Only null left

CAS: 25764-11-8 | NNd | 158.249 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 25764-11-8
Molecular Formula NNd
Molecular Weight (g/mol) 158.249
MDL Number MFCD00049990
InChI Key OVMJQLNJCSIJCH-UHFFFAOYSA-N
Synonym neodymium nitride, neodymium nitride ndn, neodymium iii nitride, neodymiumylidyneamine
PubChem CID 117628
IUPAC Name azanylidyneneodymium
SMILES N#[Nd]

Specifications

Physical Form Powder
Quantity 2 g
Formula Weight 158.25
Percent Purity 99.9%
Assay (REO)
Chemical Name or Material Neodymium(III) nitride
EINECSNumber 247-246-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only