missing translation for 'onlineSavingsMsg'
Learn More

Neodymium(III) nitride, 99.9% (REO)

Catalog No. AA4209109
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4209109 10 g
AA4209104 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4209109 Supplier Thermo Scientific Chemicals Supplier No. 04209109

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 25764-11-8 | NNd | 158.249 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 25764-11-8
Molecular Formula NNd
Molecular Weight (g/mol) 158.249
MDL Number MFCD00049990
InChI Key OVMJQLNJCSIJCH-UHFFFAOYSA-N
Synonym neodymium nitride, neodymium nitride ndn, neodymium iii nitride, neodymiumylidyneamine
PubChem CID 117628
IUPAC Name azanylidyneneodymium
SMILES N#[Nd]

Specifications

Physical Form Powder
Quantity 10 g
Formula Weight 158.25
Percent Purity 99.9%
Assay (REO)
Chemical Name or Material Neodymium(III) nitride
EINECSNumber 247-246-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only