missing translation for 'onlineSavingsMsg'
Learn More

Neocuproine hydrochloride monohydrate, For Spectrophotometric Det. of Cu, ≥99.0%, MilliporeSigma™ Supelco™

Catalog No. 111013861
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013861 1 g
111013862 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111013861 Supplier Merck Emd Millipore Supplier No. 720901G

Chemical Identifiers

MDL Number MFCD00150062
Synonym 2,9-Dimethyl-1,10-phenanthroline hydrochloride

Specifications

CAS 303136-82-5
Linear Formula C14H12N2 · HCl · H2O
Quantity 1 g
Solubility Information Water: 25 mg/mL, clear
Formula Weight 262.73
Percent Purity ≥99.0% (AT); ≥99.0%