missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neocuproine Hydrochloride Monohydrate 99.0+%, TCI America™
Specifications
Specifications
| Color | White-Yellow |
| Quantity | 1 g |
| Formula Weight | 244.72 |
| Percent Purity | ≥99.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Neocuproine Hydrochloride Monohydrate |
Safety and Handling
| TSCA | Yes |