missing translation for 'onlineSavingsMsg'
Learn More

Neocuproine Hydrochloride Monohydrate 99.0+%, TCI America™

Catalog No. N04231G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N04231G 1 g
N04235G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N04231G Supplier TCI America Supplier No. N04231G

Chemical Identifiers

CAS 303136-82-5
Molecular Formula C14H12N2
Molecular Weight (g/mol) 208.26
MDL Number MFCD00150062
InChI Key IYRGXJIJGHOCFS-UHFFFAOYSA-N
Synonym 2,9-dimethyl-1,10-phenanthroline hydrochloride monohydrate, 2,9-dimethyl-1,10-phenanthroline hydrochloride hydrate, neocuproine hydrochloride monohydrate, neocuproine hydrate hydrochloride, c14h12n2.hcl.h2o, 2,9-dimethyl-1,10-phenanthroline hydrate hydrochloride, neocuproine hydrochloride hydrate, neocuproinehydrochloridemonohydrate, neocuproine hydrochloride hydrate 98, neocuproine hydrochloride trihydrate, 98 %
PubChem CID 2723838
IUPAC Name 2,9-dimethyl-1,10-phenanthroline
SMILES CC1=CC=C2C=CC3=CC=C(C)N=C3C2=N1

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 244.72
Percent Purity ≥99.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Neocuproine Hydrochloride Monohydrate