missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neo-Clear™, (Xylene Substitute), For Microscopy, MilliporeSigma™
For Histoprocessing, Deparaffination and clearing after dehydration in staining process.
$374.84 - $1,587.52
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
M1098435000
|
Technidata
1098435000 |
5 L | Plastic Bottle |
N/A
|
|
||||
|
M1098439025
|
Technidata
1098439025 |
25 L | Barrel Pe/Met |
N/A
|
|
||||
Chemical Identifiers
| 64741-65-7 |
Specifications
| Liquid | |
| 0.78Â -Â 0.795Â g/3 (20Â °C) | |
| 6 hPa (20°C) | |
| 5 L |
| 64741-65-7 | |
| Plastic Bottle | |
| 43°C |
Safety and Handling
EINECSNumber : 265-067-2