missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NEO-823 98.0+%, TCI America™
Supplier: TCI America N1056100MG
Specifications
| NEO-823 | |
| 172°C | |
| 100 mg | |
| SXFGKYGUYHUANQ-NRJODPMSSA-N | |
| 2-[3-cyano-4-[(E)-2-[5-[(E)-2-[4-(dibutylamino)-2-phenylmethoxyphenyl]ethenyl]thiophen-2-yl]ethenyl]-5-phenyl-5-(trifluoromethyl)furan-2-ylidene]propanedinitrile | |
| 88549116 | |
| ≥98.0% (HPLC) |
| 1267603-73-5 | |
| C44H39F3N4O2S | |
| 2-[4-[2-[5-[2-(Benzyloxy)-4-(dibutylamino)styryl]thiophen-2-yl]vinyl]-3-cyano-5-phenyl-5-(trifluoromethyl)furan-2(5H)-ylidene]malononitrile, 2-[3-Cyano-4-[2-[5-[2-[4-(dibutylamino)-2-(phenylmethoxy)phenyl]ethenyl]-2-thienyl]ethenyl]-5-phenyl-5-(trifluorom | |
| CCCCN(CCCC)C1=CC(=C(C=C1)C=CC2=CC=C(S2)C=CC3=C(C(=C(C#N)C#N)OC3(C4=CC=CC=C4)C(F)(F)F)C#N)OCC5=CC=CC=C5 | |
| 744.877 | |
| 744.88 | |
| Crystalline Powder |
Chemical Identifiers
| 1267603-73-5 | |
| 744.877 | |
| 2-[4-[2-[5-[2-(Benzyloxy)-4-(dibutylamino)styryl]thiophen-2-yl]vinyl]-3-cyano-5-phenyl-5-(trifluoromethyl)furan-2(5H)-ylidene]malononitrile, 2-[3-Cyano-4-[2-[5-[2-[4-(dibutylamino)-2-(phenylmethoxy)phenyl]ethenyl]-2-thienyl]ethenyl]-5-phenyl-5-(trifluorom | |
| 2-[3-cyano-4-[(E)-2-[5-[(E)-2-[4-(dibutylamino)-2-phenylmethoxyphenyl]ethenyl]thiophen-2-yl]ethenyl]-5-phenyl-5-(trifluoromethyl)furan-2-ylidene]propanedinitrile |
| C44H39F3N4O2S | |
| SXFGKYGUYHUANQ-NRJODPMSSA-N | |
| 88549116 | |
| CCCCN(CCCC)C1=CC(=C(C=C1)C=CC2=CC=C(S2)C=CC3=C(C(=C(C#N)C#N)OC3(C4=CC=CC=C4)C(F)(F)F)C#N)OCC5=CC=CC=C5 |
Safety and Handling
TSCA : No