missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Negative Control for Rabbit Monoclonal Antibody (NCRBM/1520R) Alexa Fluor™ Plus 594, R&D Systems™
Description
Negative Control for Rabbit Monoclonal Monoclonal antibody specifically detects Negative Control for Rabbit Monoclonal in Nonspecies specific samples. It is validated for Flow Cytometry,ELISA,Immunohistochemistry,Immunoprecipitation,Western Blot,Immunohistochemistry (Paraffin),CyTOF,Immunocytochemistry/Immunofluorescence
Spécifications
Spécifications
| Antigène | Negative Control for Rabbit Monoclonal |
| Applications | Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | NCRBM/1520R |
| Conjugué | Alexa Fluor Plus 594 |
| Formule | 50mM Sodium Borate |
| Espèces hôtes | Rabbit |
| Immunogène | Recombinant full-length human Negative Control for Rabbit Monoclonal protein |
| Méthode de purification | Protein A purified |
| Quantité | 0.1 mL |
| Afficher plus de résultats |