Recombinant Protein;1MG;Human Nectin-2, Fc Tag (PV2-H5253) is expressed from human 293 cells (HEK293). It contains AA Gln 32 - Leu 360 (Accession # Q92692-2).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 62.2 kDa. The protein migrates as 70-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human PVRIG, His Tag (Cat. No. PVG-H52H4) at 2 g/mL (100 L/well) can bind Human Nectin-2, Fc Tag (Cat. No. PV2-H5253) with a linear range of 0.5-8 ng/mL (QC tested).;Immobilized Human Nectin-2, Fc Tag (Cat. No. PV2-H5253) at 10 g/mL (100 L/well) can bind Biotinylated Human TIGIT, Fc Tag, Avi Tag with a linear range of 0.156-2.5 g/mL (Routinely tested).