missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Necrosis Inhibitor, IM-54

Catalog No. AAJ64372LB0
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ64372LB0 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ64372LB0 Supplier Thermo Scientific Chemicals Supplier No. J64372LB0

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

 

A cell-permeable compound that selectively inhibits oxidative stress-induced necrosis

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 861891-50-1
Molecular Formula C19H23N3O2
Synonym 1-Methyl-3-(1-methyl-1H-indol-3-yl)-4-(pentylamino)-1H-pyrrole-2,5-dione;

Specifications

Quantity 5 mg
Description Yellow orange
Content And Storage 2°C to 8°C
For Use With (Application) A cell-permeable compound that selectively inhibits oxidative stress-induced necrosis
Formula Weight 325.40
Form Powder

RUO – Research Use Only