Learn More
New England Biolab Ltd NEBNext Microbiome DNA Enrichment Kit 24 reactions DFS Item
Supplier:  New England Biolab Ltd E2612L
The NEBNext Microbiome DNA Enrichment Kit facilitates enrichment of microbial DNA from samples containing methylated host DNA (including human), by selective binding and removal of the CpG-methylated host DNA. Importantly, microbial diversity remains intact after enrichment.
Catalog No. 501047506
Only null left
Explore available promotions