Learn More
New England Biolab Ltd NEBNext Companion Module forOxford Nanopore Technologies Ligation Sequencing 24 reactions
Catalog No. NC1671225
Encompass
This module contains the following reagents, at volumes designed for use alongside Oxford Nanopore Technologies SQK-LSK109: NEBNext FFPE DNA Repair Mix (0.048 ml) NEBNext FFPE DNA Repair Buffer (0.084 ml) NEBNext Ultra II End Prep Enzyme Mix (0.072 ml) NEBNext Ultra II End Prep Reaction Buffer (0.084 ml) Quick T4 DNA Ligase (0.240 ml) The appropriate device-specific SQK-LSK109 protocol from Oxford Nanopore Technologies should be followed for use of these reagents.
Catalog No. NC1671225 Supplier New England Biolab Ltd Supplier No. E7180S
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
This module contains the following reagents, at volumes designed for use alongside Oxford Nanopore Technologies SQK-LSK109: NEBNext FFPE DNA Repair Mix (0.048 ml) NEBNext FFPE DNA Repair Buffer (0.084 ml) NEBNext Ultra II End Prep Enzyme Mix (0.072 ml) NEBNext Ultra II End Prep Reaction Buffer (0.084 ml) Quick T4 DNA Ligase (0.240 ml) The appropriate device-specific SQK-LSK109 protocol from Oxford Nanopore Technologies should be followed for use of these reagents.