Learn More
New England Biolab Ltd NEBNext FFPE DNA Repair Mix 96 reactions
Catalog No. NC0875376
Encompass
The NEBNext FFPE DNA Repair Mix contains enzymes and buffers that are optimized to repair FFPE DNA in next generation sequencing workflows. The mix can be used upstream of any next generation sequencing library construction method, for any sequencing platform.
Catalog No. NC0875376 Supplier New England Biolab Ltd Supplier No. M6630L
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The NEBNext FFPE DNA Repair Mix contains enzymes and buffers that are optimized to repair FFPE DNA in next generation sequencing workflows. The mix can be used upstream of any next generation sequencing library construction method, for any sequencing platform.