New England Biolab Ltd NEB 5-alpha Competent E. coli (High Efficiency) 6 x 0.2 ml/tube
Catalog No. 50995054
NEB 5-alpha Competent E. coli (High Efficiency) is a derivative of the popular DH5. It is T1 phage resistant and endA deficient for high-quality plasmid preparations.
Catalog No.
50995054
Supplier
New England Biolab LtdSupplier No.
C2987I
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
NEB 5-alpha Competent E. coli (High Efficiency) is a derivative of the popular DH5. It is T1 phage resistant and endA deficient for high-quality plasmid preparations.