Learn More
New England Biolab Ltd NEB 5-alpha Competent E. coli (High Efficiency) 96 x 50 l/tube (12 x 8-tube strips)
Catalog No. 501526338
Encompass
NEB 5-alpha Competent E. coli (High Efficiency) is a derivative of the popular DH5. It is T1 phage resistant and endA deficient for high-quality plasmid preparations.
Catalog No. 501526338 Supplier New England Biolab Ltd Supplier No. C2987U
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
NEB 5-alpha Competent E. coli (High Efficiency) is a derivative of the popular DH5. It is T1 phage resistant and endA deficient for high-quality plasmid preparations.