New England Biolab Ltd NEB 10-beta Electrocompetent E.coli 6 x 0.1 ml/tube
Catalog No. 50995063
NEB 10-beta electrocompetent E. coli cells are optimized for high efficiency transformation by electroporation. These cells are ideal for DNA library constructions and all cloning purposes.
Catalog No.
50995063
Supplier
New England Biolab LtdSupplier No.
C3020K
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
NEB 10-beta electrocompetent E. coli cells are optimized for high efficiency transformation by electroporation. These cells are ideal for DNA library constructions and all cloning purposes.