New England Biolab Ltd NEB 10-beta Competent E. coli (High Efficiency) 20 x 0.05 ml/tube
Catalog No. 50995061
NEB 10-beta Competent E. coli is a derivative of the popular DH10B. It is T1 phage resistant and endonuclease I (endA1) deficient for high- quality plasmid preparations.
Catalog No.
50995061
Supplier
New England Biolab LtdSupplier No.
C3019H
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
NEB 10-beta Competent E. coli is a derivative of the popular DH10B. It is T1 phage resistant and endonuclease I (endA1) deficient for high- quality plasmid preparations.