missing translation for 'onlineSavingsMsg'
Learn More

Ndelta-Boc-Nalpha-Fmoc-L-ornithine, 96%

Catalog No. AAH6281406
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6281406 5 g
AAH6281414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6281406 Supplier Thermo Scientific Chemicals Supplier No. H6281406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 109425-55-0 | C25H30N2O6 | 454.523 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 109425-55-0
Molecular Formula C25H30N2O6
Molecular Weight (g/mol) 454.523
MDL Number MFCD00065668
InChI Key JOOIZTMAHNLNHE-NRFANRHFSA-N
Synonym fmoc-orn boc-oh, nalpha-fmoc-ndelta-boc-l-ornithine, fmoc-l-orn boc-oh, ndelta-boc-nalpha-fmoc-l-ornithine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-5-tert-butoxycarbonyl amino pentanoic acid, 2s-5-tert-butoxycarbonyl amino-2-9h-fluoren-9-ylmethoxy carbonyl amino pentanoic acid, fmoc-orn boc, pubchem10032, n-a-boc-n-deta-boc-l-ornithine, n alpha-fmoc-n delta-boc-l-ornithine
PubChem CID 2756114
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-5-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid
SMILES CC(C)(C)OC(=O)NCCCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13

Specifications

Melting Point 111°C to 115°C
Beilstein 4772025
Quantity 5 g
Formula Weight 454.52
Percent Purity 96%
Chemical Name or Material Ndelta-Boc-Nalpha-Fmoc-L-ornithine
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only