missing translation for 'onlineSavingsMsg'
Learn More

Ndelta-Boc-L-ornithine, 98%

CAS: 13650-49-2 | C10H20N2O4 | 232.28 g/mol

Supplier:  Thermo Scientific Chemicals H6277406

 View more versions of this product

Catalog No. AAH6277406


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Ndelta-Boc-L-ornithine
C10H20N2O4
h-orn boc-oh, s-2-amino-5-tert-butoxycarbonylamino-pentanoic acid, n-delta-boc-l-ornithine, boc-l-orn-oh, ndelta-boc-l-ornithine, ndelta-boc-l-ornithine, a-boc-l-ornithine, l-ornithine, n5-1,1-dimethylethoxy carbonyl, l-ornithine, n5-1,1-dimethylethoxy carbonyl, n∼5∼-tert-butoxycarbonyl-l-ornithine, n∼5∼-tert-butoxycarbonyl-l-ornithine
CC(C)(C)OC(=O)NCCCC(C(=O)O)N
5 g
6993436
98%
13650-49-2
MFCD00037219
GLZZMUULAVZVTA-ZETCQYMHSA-N
(2S)-2-amino-5-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid
232.28
232.28

Chemical Identifiers

13650-49-2
232.28
GLZZMUULAVZVTA-ZETCQYMHSA-N
6993436
CC(C)(C)OC(=O)NCCCC(C(=O)O)N
C10H20N2O4
MFCD00037219
h-orn boc-oh, s-2-amino-5-tert-butoxycarbonylamino-pentanoic acid, n-delta-boc-l-ornithine, boc-l-orn-oh, ndelta-boc-l-ornithine, ndelta-boc-l-ornithine, a-boc-l-ornithine, l-ornithine, n5-1,1-dimethylethoxy carbonyl, l-ornithine, n5-1,1-dimethylethoxy carbonyl, n∼5∼-tert-butoxycarbonyl-l-ornithine, n∼5∼-tert-butoxycarbonyl-l-ornithine
(2S)-2-amino-5-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only