missing translation for 'onlineSavingsMsg'
Learn More

NBD-F (=4-Fluoro-7-nitro-2,1,3-benzoxadiazole) 99.0+%, TCI America™

Catalog No. A5593100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A5593100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A5593100MG Supplier TCI America Supplier No. A5593100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 29270-56-2
Molecular Formula C6H2FN3O3
Molecular Weight (g/mol) 183.098
MDL Number MFCD00010196
InChI Key PGZIDERTDJHJFY-UHFFFAOYSA-N
Synonym 4-fluoro-7-nitrobenzofurazan, nbd-f, 4-fluoro-7-nitrobenzo c 1,2,5 oxadiazole, 7-fluoro-4-nitrobenzofurazan, 7-fluoro-4-nitrobenzo-2-oxa-1,3-diazole, 4-fluoro-7-nitrobenz-2-oxa-1,3-diazole, 2,1,3-benzoxadiazole, 4-fluoro-7-nitro, nbd-fluoride, nbd-f cpd, 4-fluoro-7-nitrobenzo c 1,2,5-oxadiazole
PubChem CID 122123
IUPAC Name 4-fluoro-7-nitro-2,1,3-benzoxadiazole
SMILES C1=C(C2=NON=C2C(=C1)F)[N+](=O)[O-]

Specifications

Melting Point 55°C
Quantity 100 mg
Formula Weight 183.10
Percent Purity ≥99.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material NBD-F (=4-Fluoro-7-nitro-2,1,3-benzoxadiazole) [for HPLC Labeling]
TSCA No
Recommended Storage Freezer