missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NBD-556 98.0+%, TCI America™
$438.05 - $1204.17
Chemical Identifiers
| CAS | 333353-44-9 |
|---|---|
| Molecular Formula | C17H24ClN3O2 |
| Molecular Weight (g/mol) | 337.848 |
| MDL Number | MFCD00469538 |
| InChI Key | ZKXLQCIOURANAD-UHFFFAOYSA-N |
| Synonym | N1-(4-Chlorophenyl)-N2-(2,2,6,6-tetramethyl-4-piperidinyl)ethanediamide, N1-(4-Chlorophenyl)-N2-(2,2,6,6-tetramethylpiperidin-4-yl)oxalamide |
| PubChem CID | 1570601 |
| IUPAC Name | N'-(4-chlorophenyl)-N-(2,2,6,6-tetramethylpiperidin-4-yl)oxamide |
| SMILES | CC1(CC(CC(N1)(C)C)NC(=O)C(=O)NC2=CC=C(C=C2)Cl)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
N109025MG
|
TCI America
N109025MG |
25 mg |
Each for $438.05
|
|
|||||
|
N1090100MG
|
TCI America
N1090100MG |
100 mg |
Each for $1,204.17
|
|
|||||
Chemical Identifiers
| 333353-44-9 | |
| 337.848 | |
| ZKXLQCIOURANAD-UHFFFAOYSA-N | |
| 1570601 | |
| CC1(CC(CC(N1)(C)C)NC(=O)C(=O)NC2=CC=C(C=C2)Cl)C |
| C17H24ClN3O2 | |
| MFCD00469538 | |
| N1-(4-Chlorophenyl)-N2-(2,2,6,6-tetramethyl-4-piperidinyl)ethanediamide, N1-(4-Chlorophenyl)-N2-(2,2,6,6-tetramethylpiperidin-4-yl)oxalamide | |
| N'-(4-chlorophenyl)-N-(2,2,6,6-tetramethylpiperidin-4-yl)oxamide |
Specifications
| 333353-44-9 | |
| MFCD00469538 | |
| 2811 | |
| ZKXLQCIOURANAD-UHFFFAOYSA-N | |
| N'-(4-chlorophenyl)-N-(2,2,6,6-tetramethylpiperidin-4-yl)oxamide | |
| 1570601 | |
| ≥98.0% (HPLC) | |
| NBD-556 |
| C17H24ClN3O2 | |
| 25 mg | |
| N1-(4-Chlorophenyl)-N2-(2,2,6,6-tetramethyl-4-piperidinyl)ethanediamide, N1-(4-Chlorophenyl)-N2-(2,2,6,6-tetramethylpiperidin-4-yl)oxalamide | |
| CC1(CC(CC(N1)(C)C)NC(=O)C(=O)NC2=CC=C(C=C2)Cl)C | |
| 337.848 | |
| 337.85 | |
| Crystalline Powder |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator