missing translation for 'onlineSavingsMsg'
Learn More
Learn More
International Enviroguard™ Navy Blue Polypropylene Coveralls with Hood and Boots
Description
- Polypropylene coveralls comfortably fit over work clothes and protect against non-hazardous materials like dust, dirt, and grime
- Lightweight coverall offers a breathable alternative to traditional, bulky coverall suits
- Elastic wrists provide a secure fit to keep sleeves in place and decrease the possibility of skin exposure
- Elastic wrists, open ankles
- Disposable polypropylene coveralls are a great option for preventing cross-contamination of products, while also keeping you safe from particulates
- Stay cool, comfortable, and protected from head-to-toe
- Possible applications include food processing, construction, pharmaceuticals, environmental cleanup, and more
Specifications
Specifications
| Product Type | Light-duty Coveralls |
| Size | 2X-Large |
| Gender | Unisex |
| Material | Polypropylene |
| Color | Navy |
| Disposable | Single-use |
| Waist Style | Standard |
| Ankle Style | Attached Boots |
| Cuff Style | Elastic |
| Neck Style | Hooded |
| Show More |