missing translation for 'onlineSavingsMsg'
Learn More
Learn More
International Enviroguard™ Navy Blue Polypropylene Coveralls with Elastic Wrists and Open Ankles
Disposable coveralls provide low-cost protection against dust, dirt, and grime.
Supplier: International Enviroguard™ N20124XL
Description
- Polypropylene coveralls comfortably fit over work clothes and protect against non-hazardous materials like dust, dirt, and grime
- Lightweight coverall offers a breathable alternative to traditional, bulky coverall suits
- Elastic wrists provide a secure fit to keep sleeves in place and decrease the possibility of skin exposure
- Elastic wrists, open ankles
- Disposable polypropylene coveralls are a great option for preventing cross-contamination of products, while also keeping you safe from particulates
- Stay cool, comfortable, and protected from head-to-toe
- Possible applications include food processing, construction, pharmaceuticals, environmental cleanup, and more
Specifications
| Light-duty Coveralls | |
| Unisex | |
| Navy | |
| Standard | |
| Elastic | |
| Standard Collar | |
| Serged | |
| Particle, Dust | |
| N2012 |
| 4X-Large | |
| Polypropylene | |
| Single-use | |
| Open | |
| Collared | |
| Front Zipper | |
| Non-sterile | |
| Yes | |
| None |