Learn More
International Enviroguard™ Navy Blue Polypropylene Coveralls with Elastic Wrists and Open Ankles
Disposable coveralls provide low-cost protection against dust, dirt, and grime.
Supplier:  International Enviroguard™ N20125XL
 View more versions of this product
Catalog No. 17-183-969
Only null left
Explore available promotions
Description
Description
  • Polypropylene coveralls comfortably fit over work clothes and protect against non-hazardous materials like dust, dirt, and grime
  • Lightweight coverall offers a breathable alternative to traditional, bulky coverall suits
  • Elastic wrists provide a secure fit to keep sleeves in place and decrease the possibility of skin exposure
  • Elastic wrists, open ankles
  • Disposable polypropylene coveralls are a great option for preventing cross-contamination of products, while also keeping you safe from particulates
  • Stay cool, comfortable, and protected from head-to-toe
  • Possible applications include food processing, construction, pharmaceuticals, environmental cleanup, and more
Specifications
Specifications
Light-duty Coveralls
Unisex
Navy
Standard
Elastic
Standard Collar
Serged
Particle, Dust
N2012
5X-Large
Polypropylene
Single-use
Open
Collared
Front Zipper
Non-sterile
Yes
None
Product Suggestions
Product Suggestions