missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Nav1.5 Peptide
Specifications
Specifications
| Gene ID (Entrez) | 6331 |
| Species | Human |
| Product Line | Novus |
| Content And Storage | Store at -80°C. Avoid freeze/thaw cycles. |
| For Use With (Application) | This Antibody is Useful for Peptode ELISA and Western Blot. Preliminary Experiments in Ysates of Human Breast and Heart Tissues Gave no Specific Signal But Low Background. |
| Gene Alias | cardiac sodium channel alpha subunit, cardiac tetrodotoxin-insensitive voltage-dependent sodium channel alpha subunit, CDCD2VF1, HB1sodium channel, voltage-gated, type V, alpha (long QT syndrome 3), HB2, HBBD, HH1CMD1E, ICCD, IVF, LQT3SSS1CMPD2, Nav1.5, PFHB1, Sodium channel protein cardiac muscle subunit alpha, sodium channel protein type 5 subunit alpha, sodium channel protein type V alpha subunit, Sodium channel protein type V subunit alpha, sodium channel, voltage-gated, type V, alpha subunit, Voltage-gated sodium channel subunit alpha Nav1.5 |
| Gene Symbol | SCN5A |
| Quantity | 0.1 mg |
| Research Category | Cancer, Hypoxia |
| Storage Buffer | Peptide dissolved in dH2O. Contains no BSA. |