missing translation for 'onlineSavingsMsg'
Learn More

GOJO™ NATURAL ORANGE™ Smooth Hand Cleaner

Catalog No. 19044104
Change view
Click to view available options
Volume (Metric):
2000 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
19044104 2000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19044104 Supplier GOJO™ Supplier No. 725004

Quick-acting, lotion hand cleaner.

  • Refill for GOJO™ PRO™ TDX™ Push-Style Dispenser
  • Contains a natural citrus ingredient
  • Cleans without harsh solvents
  • SANITARY SEALED™ refill helps prevent contamination
  • Fresh dispensing valve with each refill
  • Effective on dirt and oil

Specifications

Product Type Hand Cleanser
Color White
Form Gritty Liquid
Size 2000 mL
Volume (Metric) 2000 mL