Native E. coli beta Galactosidase protein is a Escherichia coli K-12 Full Length protein, in the 2 to 1024 aa range, expressed in Escherichia coli, with >95% purity and suitable for SDS-PAGE.
Protein/ Peptide/ Enzyme Synonyms: b0344, JW0335, lacZ, Beta-galactosidase, Beta-gal, Lactase
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement