Learn More
Abcam Inc Native Cow Factor IX/PTC protein, 100UG
Catalog No. 502710218
Encompass
Native Cow Factor IX/PTC protein is a Cow Full Length protein, expressed in Native, with >95% purity and suitable for SDS-PAGE, FuncS.
Protein/ Peptide/ Enzyme Synonyms: Coagulation factor IX, Christmas factor
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 502710218 Supplier Abcam Inc Supplier No. AB62547100UG
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Native Cow Factor IX/PTC protein is a Cow Full Length protein, expressed in Native, with >95% purity and suitable for SDS-PAGE, FuncS.
Protein/ Peptide/ Enzyme Synonyms: Coagulation factor IX, Christmas factor
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement